Merck
  • A folding-based approach for the luminescent detection of a short RNA hairpin.
A folding-based approach for the luminescent detection of a short RNA hairpin.
Journal of the American Chemical Society (2013-02-26)
Cristina Penas, Elena Pazos, José L Mascareñas, M Eugenio Vázquez
ABSTRACT
We report the rational design of a 20-mer basic peptide, derived from the transcriptional antitermination protein N of bacteriophage P22, equipped with a luminescent DOTA[Tb(3+)] macrocyclic complex and a sensitizing tryptophan antenna. Folding of this peptide into an α helical conformation, which occurs upon binding to its target boxB RNA hairpin, results in a large increase in luminescence emission. Therefore, the peptide construct works as a highly sensitive and selective probe for this RNA hairpin.