Merck
  • Ginkgo biloba extract and long-term cognitive decline: a 20-year follow-up population-based study.
Ginkgo biloba extract and long-term cognitive decline: a 20-year follow-up population-based study.
PloS one (2013-01-18)
Hélène Amieva, Céline Meillon, Catherine Helmer, Pascale Barberger-Gateau, Jean François Dartigues
ABSTRACT
Numerous studies have looked at the potential benefits of various nootropic drugs such as Ginkgo biloba extract (EGb761®; Tanakan®) and piracetam (Nootropyl®) on age-related cognitive decline often leading to inconclusive results due to small sample sizes or insufficient follow-up duration. The present study assesses the association between intake of EGb761® and cognitive function of elderly adults over a 20-year period. The data were gathered from the prospective community-based cohort study 'Paquid'. Within the study sample of 3612 non-demented participants aged 65 and over at baseline, three groups were compared: 589 subjects reporting use of EGb761® at at least one of the ten assessment visits, 149 subjects reporting use of piracetam at one of the assessment visits and 2874 subjects not reporting use of either EGb761® or piracetam. Decline on MMSE, verbal fluency and visual memory over the 20-year follow-up was analysed with a multivariate mixed linear effects model. A significant difference in MMSE decline over the 20-year follow-up was observed in the EGb761® and piracetam treatment groups compared to the 'neither treatment' group. These effects were in opposite directions: the EGb761® group declined less rapidly than the 'neither treatment' group, whereas the piracetam group declined more rapidly (β = -0.6). Regarding verbal fluency and visual memory, no difference was observed between the EGb761® group and the 'neither treatment' group (respectively, β = 0.21 and β = -0.03), whereas the piracetam group declined more rapidly (respectively, β = -1.40 and β = -0.44). When comparing the EGb761® and piracetam groups directly, a different decline was observed for the three tests (respectively β = -1.07, β = -1.61 and β = -0.41). Cognitive decline in a non-demented elderly population was lower in subjects who reported using EGb761® than in those who did not. This effect may be a specific medication effect of EGb761®, since it was not observed for another nootropic medication, piracetam.