Merck
  • Affinity labeling of hepatitis C virus replicase with a nucleotide analogue: identification of binding site.
Affinity labeling of hepatitis C virus replicase with a nucleotide analogue: identification of binding site.
Biochemistry (2012-12-28)
Dinesh Manvar, Kamlendra Singh, Virendra N Pandey
ABSTRACT
We have used an ATP analogue 5'-[p-(fluorosulfonyl)benzoyl]adenosine (FSBA) to modify HCV replicase in order to identify the ATP binding site in the enzyme. FSBA inactivates HCV replicase activity in a concentration-dependent manner with a binding stoichiometry of 2 moles of FSBA per mole of enzyme. The enzyme activity is protected from FSBA in the presence of rNTP substrates or double-stranded RNA template primers that do not support ATP as the incoming nucleotide but not in the presence of polyrU.rA(26). HPLC analysis of tryptic peptides of FSBA-modified enzyme revealed the presence of two distinct peptides eluted at 23 and 36 min; these were absent in the control. Further we noted that both peptides were protected from FSBA modification in the presence of Mg·ATP. The LC/MS/MS analysis of the affinity-labeled tryptic peptides purified from HPLC, identified two major modification sites at positions 382 (Tyr), and 491 (Lys) and a minor site at position 38 (Tyr). To validate the functional significance of Tyr38, Tyr382, and Lys491 in catalysis, we individually substituted these residues by alanine and examined their ability to catalyze RdRp activity. We found that both Y382A and K491A mutants were significantly affected in their ability to catalyze RdRp activity while Y38A remained unaffected. We further observed that both Y382A and K491A mutants were not affected in their ability to bind template primer but were significantly affected in their ability to photo-cross-link ATP in the absence or presence of template primer.