Merck
  • Comparison of five methods for oxacillin susceptibility testing of Staphylococcus aureus isolates from cystic fibrosis patients.
Comparison of five methods for oxacillin susceptibility testing of Staphylococcus aureus isolates from cystic fibrosis patients.
Revista do Instituto de Medicina Tropical de Sao Paulo (2012-11-16)
Marcelo J Mimica, Rozane L Carvalho, Eitan N Berezin, Neiva Damaceno, Hélio H Caiaffa-Filho
ABSTRACT
Methicillin-resistant Staphylococcus aureus (MRSA) are now a worldwide problem. Cystic fibrosis (CF) patients are commonly colonized and infected by MRSA. Accurate oxacillin susceptibility testing is mandatory for the adequate management of these patients. We performed a comparison of the accuracy of different tests in CF isolates, including methicillin-susceptible S. aureus and MRSA with different SCCmec types, and using the mecA gene as the gold-standard. The sensitivity and specificity of oxacillin disc, Etest, and oxacillin agar screening plate were 100%. Sensitivity of the cefoxitin disc was 85% and specificity was 100%. For clinically relevant isolates, laboratories may consider the use of a combination of two phenotypic methods.
Product Description
Supelco
Oxacillin sodium salt monohydrate, VETRANAL®, analytical standard