Merck
  • Rapid photogeneration of silver nanoparticles in ethanolic solution: a kinetic study.
Rapid photogeneration of silver nanoparticles in ethanolic solution: a kinetic study.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-11-06)
Bahareh Yahyaei, Saeid Azizian
ABSTRACT
Ag nanoparticles have been synthesized via UV irradiation of ethanolic solution of AgNO3 in presence of pluronic F127 surfactant. This study is aimed at developing a rapid, simple and green method to prepare Ag nanoparticles and understanding its generation kinetics. The formation dependency of silver nanoparticles on the concentration of reactants, UV exposure time and temperature has been investigated by using UV-vis spectroscopy. The 2D map technique has been used for the first time to estimate the switching time between the nucleation and growth of Ag nanoparticles. Appropriate kinetic models were used for modelling of both stages.