Merck
  • Clinical effects of psychological intervention and drug therapy against peptic ulcer.
Clinical effects of psychological intervention and drug therapy against peptic ulcer.
Asian Pacific journal of tropical medicine (2012-10-10)
Duo-Yu Wu, Min Guo, Yun-Suo Gao, Yan-Hai Kang, Jun-Cheng Guo, Xiang-Ling Jiang, Feng Chen, Tao Liu, Min Li
ABSTRACT
To evaluate the clinical effects of psychological interventions and drug therapy against peptic ulcer. 96 patients with peptic ulcer were divided into control group with Tagamet 800 mg per evening p.o. and trial group with psychological intervention on the basis of drug treatment. There were significant differences between the two groups (P<0.05), the trial group showed that the anxiety and depression cases declined obviously and effective rate of ulcer therapy was much higher than control group. In sum, psychological intervention combined with drug therapy provides an effective method for ulcer treatment.