Merck
  • Development of an enzyme-linked immunosorbent assay for determination of the furaltadone etabolite, 3-amino-5-morpholinomethyl-2-oxazolidinone (AMOZ) in animal tissues.
Development of an enzyme-linked immunosorbent assay for determination of the furaltadone etabolite, 3-amino-5-morpholinomethyl-2-oxazolidinone (AMOZ) in animal tissues.
Biomedical and environmental sciences : BES (2012-10-03)
Peng Jie Luo, Wen Xiao Jiang, Ross C Beier, Jian Zhong Shen, Hai Yang Jiang, Hong Miao, Yun Feng Zhao, Xia Chen, Yong Ning Wu
ABSTRACT
To determine 3-amino-5-morpholinomethyl-2-oxazolidinone (AMOZ) residues released from protein bound AMOZ in animal tissues. Polyclonal and monoclonal antibodies were produced in this study. A rapid, sensitive, and specific competitive direct enzyme-linked immunosorbent assay (cdELISA) was developed. Rabbit polyclonal antibodies were used in the optimized cdELISA method, and exhibited negligible cross-reactivity with other compounds structurally related to AMOZ. The IC(50) of the polyclonal antibody was 0.16 ng/mL. The method limit of detection in four different types of animal and fish tissues was less than 0.06 μg/kg. Recoveries ranged from 80% to 120% for fortified samples with the coefficient of variation values less than 15%. The results of the cdELISA method were in good agreement with the results from an established liquid chromatography-tandem mass spectrometry confirmatory method used for AMOZ residues. The cdELISA method developed in the present study is a convenient practical tool for screening large numbers of animal and fish tissue samples for the the detection of released protein bound AMOZ residues.