Merck
  • Long-term immunogenicity of two doses of 2009 A/H1N1v vaccine with and without AS03(A) adjuvant in HIV-1-infected adults.
Long-term immunogenicity of two doses of 2009 A/H1N1v vaccine with and without AS03(A) adjuvant in HIV-1-infected adults.
AIDS (London, England) (2012-09-29)
Christine Durier, Corinne Desaint, Frédéric Lucht, Pierre-Marie Girard, Yves Lévy, Thierry May, Christian Michelet, Agathe Rami, François Roman, Jean-François Delfraissy, Jean-Pierre Aboulker, Odile Launay
ABSTRACT
In immunocompromised patients, alternative schedules more immunogenic than the standard influenza vaccine regimen are necessary to enhance and prolong vaccine efficacy. We previously reported that the AS03A-adjuvanted 2009 A/H1N1v vaccine yielded a higher short-term immune response than the nonadjuvanted one in HIV-1-infected adults. This study reports the long-term persistence of the immune response. In a prospective, multicenter, randomized, patient-blinded trial, two doses of AS03A-adjuvanted H1N1v vaccine containing 3.75 μg haemagglutinin (n = 155; group A) or nonadjuvanted H1N1v vaccine containing 15 μg haemagglutinin (n = 151; group B), were administered 21 days apart. Haemagglutination inhibition and neutralizing antibodies were assessed 6 and 12 months after vaccination. In group A and B, the seroprotection rates were 83.7 and 59.4% at month 6, and 70.4 and 49.3 at month 12, respectively. In a multivariate analysis, persistence of seroprotection 12 months after vaccination was negatively associated with current smoking (odds ratio = 0.6, P = 0.03) and positively related with the AS03A-adjuvanted H1N1v vaccine (odds ratio = 2.7, P = 0.0002). In HIV-1-infected adults, two doses of adjuvanted influenza vaccine induce long-term persistence of immune response up to 1 year after vaccination.
Sigma-Aldrich
Squalene, ≥98%, liquid