Merck
  • Simple and inexpensive incorporation of 19F-tryptophan for protein NMR spectroscopy.
Simple and inexpensive incorporation of 19F-tryptophan for protein NMR spectroscopy.
Chemical communications (Cambridge, England) (2012-09-25)
Peter B Crowley, Ciara Kyne, William B Monteith
ABSTRACT
Fluorine-containing amino acids are valuable probes for the biophysical characterization of proteins. Current methods for (19)F-labeled protein production involve time-consuming genetic manipulation, compromised expression systems and expensive reagents. We show that Escherichia coli BL21, the workhorse of protein production, can utilise fluoroindole for the biosynthesis of proteins containing (19)F-tryptophan.