Merck
  • Evaluation of fluocinolone acetonide sustained release implant (Retisert) dissociation during implant removal and exchange surgery.
Evaluation of fluocinolone acetonide sustained release implant (Retisert) dissociation during implant removal and exchange surgery.
American journal of ophthalmology (2012-09-18)
Benjamin P Nicholson, Rishi P Singh, Jonathan E Sears, Careen Y Lowder, Peter K Kaiser
ABSTRACT
To characterize the phenomenon of dissociation of the 2 components of the sustained-release fluocinolone acetonide implant (Retisert; Bausch & Lomb) during removal or exchange procedures, or both, and to evaluate outcomes after these events. Retrospective, observational case series. Retrospective review of 27 consecutive sustained-release fluocinolone acetonide implant (Retisert) exchanges or removals between 2001 and 2010 at the Cole Eye Institute. All patients had received the implant as treatment for noninfectious uveitis. Preoperative and postoperative characteristics were analyzed, and operative reports were reviewed to characterize the effects of intraoperative implant dissociation. Twenty-seven sustained-release fluocinolone acetonide implant (Retisert) exchange or removal surgeries were performed by 3 surgeons in 20 eyes of 19 patients. Of these 27 procedures, dissociation of the implant strut from the drug-containing cup occurred in 11 eyes (40.7%). Retrieval of the dislocated cup led to intraoperative complications, including posterior retinal tear (n = 1) and limited suprachoroidal hemorrhage (n = 1). The length of time that the implant resided in the eye correlated significantly with cup dissociation. Dissociated implants resided a mean of 47.4 months, whereas intact implants resided a mean of 32.5 months (P = .0032). There was no long-term or short-term vision loss attributed to intraoperative implant dissociation. Sustained-release fluocinolone acetonide implant (Retisert) dissociation is a common occurrence in exchange or removal procedures, or both. The longer an implant resided in the eye, the greater the tendency toward dissociation. Preparation for this complication should be contemplated in any implant removal or exchange procedure.
Product Description
Fluocinolone acetonide, European Pharmacopoeia (EP) Reference Standard