Merck
  • Clinical efficacy of antazoline in rapid cardioversion of paroxysmal atrial fibrillation--a protocol of a single center, randomized, double-blind, placebo-controlled study (the AnPAF Study).
Clinical efficacy of antazoline in rapid cardioversion of paroxysmal atrial fibrillation--a protocol of a single center, randomized, double-blind, placebo-controlled study (the AnPAF Study).
Trials (2012-09-13)
Michal M Farkowski, Aleksander Maciag, Rafal Dabrowski, Mariusz Pytkowski, Ilona Kowalik, Hanna Szwed
ABSTRACT
Rapid conversion of atrial fibrillation (AF) to sinus rhythm may be achieved by the administration of class IA, IC and III antiarrhythmic drugs or vernakalant hydrochloride. However, that treatment may be related to potential pro-arrhythmia, lack of efficacy or the exceptionally high cost of a compound used. Antazoline is a first generation antihistaminic agent with chinidin-like properties. When administered intravenously, antazoline exerts a strong antiarrhythmic effect on supraventricular arrhythmia, especially on AF, facilitating rapid conversion to sinus rhythm. Despite a relative lack of published data antazoline has been marketed in Poland and widely used in cardiology wards and emergency rooms for many years due to its efficacy, safety and rapid onset of action within minutes of administration. A randomized, double blind, placebo-controlled, superiority clinical trial was designed to assess clinical efficacy of antazoline in rapid conversion of AF to sinus rhythm. Eligible patients will present AF lasting less than 43 hours, will be in stable cardio-pulmonary condition and will have no prior history of advanced heart failure or significant valvular disease. Long-term antiarrhythmic therapy is not considered an exclusion criterion. Subjects who fulfill selection criteria will be randomly assigned to receive intravenously either antazoline or placebo in divided doses and observed for 1.5 hours after conversion to sinus rhythm or after the last i.v. bolus. Primary end point will be the conversion of AF to sinus rhythm confirmed in an electrocardiogram (ECG) during the observation period. Secondary end points will be comprised of time to conversion and return of AF during the observation period. Special consideration will be given to the observation of any adverse events. A sample size of 80 patients was calculated based on the following assumptions: two-tailed test, a type I error of 0.01, a power of 90%, efficacy of placebo 5%, efficacy of antazoline 50% and 20% drop-out rate to fulfill the criteria of intention-to-treat analysis. Due to the presumed lack of statistical power, the secondary end points and safety endpoints will be considered exploratory. ClinicalTrials.gov, NCT01527279.