Merck
  • [Effect of a combination of glutamyl-tryptophan and glycyrrhizic acid on the course of acute infection caused by influenza (H3H2) virus in mice].
[Effect of a combination of glutamyl-tryptophan and glycyrrhizic acid on the course of acute infection caused by influenza (H3H2) virus in mice].
Voprosy virusologii (2012-08-22)
V S Smirnov, V V Zarubaev, P M Anfimov, A A Shtro
ABSTRACT
The purpose of the study was to evaluate the modulating effect of glutamyl-tryptophan (EW), glycyrrhizic acid (GA), and their combination on the course of experimental infection caused by influenza A (H3N2) virus in mice. The animals were infected with influenza A/Aichi/2/68 (H3N2) virus in a dose of 1 or 10 LD50. GA (10 mg/kg body weight) and EW (0.1, 10, and 1000 microg/kg) alone or in combination were intraperitoneally injected for 5 days, starting on day 1 of virus infection. Rimantadine 50 mg/kg/day was used as a comparison drug. The combination of EW (1000 microg/kg) and GA (10 mg/kg) was ascertained to exert the maximum protective effect manifesting itself in reducing the death of infected animals (by 75-79% compared to the control depending on the viral dose) and the titers of viruses accumulated in the lung (5-6 log EID50) and in preventing lung tissue edema and inflammation. The noted effect was comparable with that seen in the use of rimantadine. The agents used alone had a lower efficacy than rimantadine. The findings permit the combination of GA and EW to be considered to be a promising agent for the treatment of influenza.