Merck
  • Asymmetric construction of spirocyclohexanonerhodanines catalyzed by simple diamine derived from chiral tert-leucine.
Asymmetric construction of spirocyclohexanonerhodanines catalyzed by simple diamine derived from chiral tert-leucine.
Chemical communications (Cambridge, England) (2012-08-07)
Wenbin Wu, Huicai Huang, Xiaoqian Yuan, Kailong Zhu, Jinxing Ye
ABSTRACT
A diamine-catalyzed asymmetric tandem reaction between α,β-unsaturated ketones and rhodanine derivatives has been developed to synthesize various spirocyclic compounds with high stereoselectivities (up to 99% ee and >20:1 dr). The products obtained contain two pharmaceutically relevant features: the biologically active rhodanine moiety embedded in a spirocyclic unit.
Sigma-Aldrich
Rhodanine, 97%