- Electrochemical detection for dynamic analyses of a redox component in droplets using a local redox cycling-based electrochemical (LRC-EC) chip device.
Electrochemical detection for dynamic analyses of a redox component in droplets using a local redox cycling-based electrochemical (LRC-EC) chip device.
Chemical communications (Cambridge, England) (2012-07-20)
Kosuke Ino, Yusuke Kanno, Taku Nishijo, Takehito Goto, Toshiharu Arai, Yasufumi Takahashi, Hitoshi Shiku, Tomokazu Matsue
PMID22810361
ABSTRACT
This report describes the electrochemical detection of a redox component in droplets using a local redox cycling-based electrochemical (LRC-EC) chip device consisting of 256 sensors. The time-course analyses showed that the redox compound in the droplet was dynamically changed during droplet evaporation or mass transfer through a water/oil interface.