Merck
  • A tandem Heck-aza-Michael addition protocol for the one-pot synthesis of isoindolines from unprotected amines.
A tandem Heck-aza-Michael addition protocol for the one-pot synthesis of isoindolines from unprotected amines.
Organic & biomolecular chemistry (2012-07-17)
Ke Chen, Sumod A Pullarkat
ABSTRACT
A palladacycle-catalyzed tandem Heck-intramolecular aza-Michael reaction protocol has been developed for the one-pot synthesis of 1-substituted isoindolines from N-unprotected 2-bromobenzylamines and acrylates with high yields.