Merck
The effects of hormonal contraceptives on female sexuality: a review.
The journal of sexual medicine (2012-07-14)
Lara J Burrows, Maureen Basha, Andrew T Goldstein
ABSTRACT
Hormonal contraceptives can influence female sexual function. The goal of this article was to provide a comprehensive review of the effects that various hormonal contraceptives may have on female sexual function. A Medline search was conducted using several terms related to and including the terms contraception, oral contraceptive, female sexual function, dyspareunia, libido, and sexual desire. A thorough review of the effects of hormonal contraceptives on female sexual function. The sexual side effects of hormonal contraceptives are not well studied, particularly with regard to impact on libido. There appears to be mixed effects on libido, with a small percentage of women experiencing an increase or a decrease, and the majority being unaffected. Healthcare providers must be aware that hormonal contraceptive can have negative effects on female sexuality so they can counsel and care for their patients appropriately.