Merck
  • Effects of blood pressure lowering on cardiovascular outcomes in different cardiovascular risk groups among participants with type 2 diabetes.
Effects of blood pressure lowering on cardiovascular outcomes in different cardiovascular risk groups among participants with type 2 diabetes.
Diabetes research and clinical practice (2012-06-09)
Susan van Dieren, Andre P Kengne, John Chalmers, Joline W J Beulens, Mark E Cooper, Diederick E Grobbee, Stephen Harrap, Giuseppe Mancia, Bruce Neal, Anushka Patel, Neil Poulter, Yvonne T van der Schouw, Mark Woodward, Sophia Zoungas
ABSTRACT
To asses differences in treatment effects of a fixed combination of perindopril-indapamide on major clinical outcomes in patients with type 2 diabetes across subgroups of cardiovascular risk. 11,140 participants with type 2 diabetes, from the ADVANCE trial, were randomized to perindopril-indapamide or matching placebo. The Framingham equation was used to calculate 5-year CVD risk and to divide participants into two risk groups, moderate-high risk (<25% and no history of macrovascular disease), very high risk (>25% and/or history of macrovascular disease). Endpoints were macrovascular and microvascular events. The mean age of participants was 66 years (42.5% female). 1000 macrovascular and 916 microvascular events were recorded over follow-up of 4.3 years. Relative treatment effects were similar across risk groups, (all P-values for heterogeneity ≥0.38). Hazard ratios for combined macro- and microvascular events were 0.89 (0.77-1.03) for the moderate-high risk and 0.92 (0.81-1.03) for the very high risk. Absolute treatment effects tended to be greater in the high risk groups although differences were not statistically significant (P>0.05). Relative effects of blood pressure lowering with perindopril-indapamide on cardiovascular outcomes were similar across risk groups whilst absolute effects trended to be greater in the high risk group.