Merck
  • Stenotrophomonas maltophilia infections in a general hospital: patient characteristics, antimicrobial susceptibility, and treatment outcome.
Stenotrophomonas maltophilia infections in a general hospital: patient characteristics, antimicrobial susceptibility, and treatment outcome.
PloS one (2012-05-25)
George Samonis, Drosos E Karageorgopoulos, Sofia Maraki, Panagiotis Levis, Dimitra Dimopoulou, Nikolaos A Spernovasilis, Diamantis P Kofteridis, Matthew E Falagas
ABSTRACT
Stenotrophomonas maltophilia is acquiring increasing importance as a nosocomial pathogen. We retrospectively studied the characteristics and outcome of patients with any type of S. maltophilia infection at the University Hospital of Heraklion, Crete, Greece, between 1/2005-12/2010. S. maltophilia antimicrobial susceptibility was tested with the agar dilution method. Prognostic factors for all-cause in-hospital mortality were assessed with multivariate logistic regression. Sixty-eight patients (median age: 70.5 years; 64.7% males) with S. maltophilia infection, not related to cystic fibrosis, were included. The 68 patients were hospitalized in medical (29.4%), surgical (26.5%), hematology/oncology departments (23.5%), or the intensive care units (ICU; 20.6%). The most frequent infection types were respiratory tract (54.4%), bloodstream (16.2%), skin/soft tissue (10.3%), and intra-abdominal (8.8%) infection. The S. maltophilia-associated infection was polymicrobial in 33.8% of the cases. In vitro susceptibility was higher to colistin (91.2%), trimethoprim/sulfamethoxazole and netilmicin (85.3% each), and ciprofloxacin (82.4%). The empirical and the targeted treatment regimens were microbiologically appropriate for 47.3% and 63.6% of the 55 patients with data available, respectively. Most patients received targeted therapy with a combination of agents other than trimethoprim/sulfamethoxazole. The crude mortality and the mortality and the S. maltophilia infection-related mortality were 14.7% and 4.4%, respectively. ICU hospitalization was the only independent prognostic factor for mortality. S. maltophilia infection in a general hospital can be associated with a good prognosis, except for the patients hospitalized in the ICU. Combination reigmens with fluoroquinolones, colistin, or tigecycline could be alternative treatment options to trimethoprim/sulfamethoxazole.