Merck
  • Phthalide and isocoumarin derivatives produced by an Acremonium sp. isolated from a mangrove Rhizophora apiculata.
Phthalide and isocoumarin derivatives produced by an Acremonium sp. isolated from a mangrove Rhizophora apiculata.
Journal of natural products (2012-04-25)
Vatcharin Rukachaisirikul, Aekkachai Rodglin, Yaowapa Sukpondma, Souwalak Phongpaichit, Jirayu Buatong, Jariya Sakayaroj
ABSTRACT
Nine new fungal metabolites, one phthalide derivative, acremonide (1), and eight isocoumarin derivatives, acremonones A-H (2-9), were isolated from the mangrove-derived fungus Acremonium sp. PSU-MA70 together with 10 known compounds. Their structures were determined by NMR analysis. The known 8-deoxytrichothecin and trichodermol exhibited moderate antifungal activity against Candida albicans and Cryptococcus neoformanns, respectively.