Merck
  • 2-Aminopyrimidine based 4-aminoquinoline anti-plasmodial agents. Synthesis, biological activity, structure-activity relationship and mode of action studies.
2-Aminopyrimidine based 4-aminoquinoline anti-plasmodial agents. Synthesis, biological activity, structure-activity relationship and mode of action studies.
European journal of medicinal chemistry (2012-03-31)
Kamaljit Singh, Hardeep Kaur, Kelly Chibale, Jan Balzarini, Susan Little, Prasad V Bharatam
ABSTRACT
2-Aminopyrimidine based 4-aminoquinolines were synthesized using an efficacious protocol. Some of the compounds showed in vitro anti-plasmodial activity against drug-sensitive CQ(S) (3D7) and drug-resistant CQ(R) (K1) strains of Plasmodium falciparum in the nM range. In particular, 5-isopropyloxycarbonyl-6-methyl-4-(2-nitrophenyl)-2-[(7-chloroquinolin-4-ylamino)butylamino] pyrimidine depicted the lowest IC(50) (3.6 nM) value (56-fold less than CQ) against CQ(R) strain. Structure-activity profile and binding with heme, μ-oxo-heme have been studied. Binding assays with DNA revealed better binding with target parasite type AT rich pUC18 DNA. Most compounds were somewhat cytotoxic, but especially cytostatic. Molecular docking analysis with Pf DHFR allowed identification of stabilizing interactions.
Sigma-Aldrich
2-Aminopyrimidine, 97%