- A fungal ketoreductase domain that displays substrate-dependent stereospecificity.
A fungal ketoreductase domain that displays substrate-dependent stereospecificity.
Nature chemical biology (2012-03-13)
Hui Zhou, Zhizeng Gao, Kangjian Qiao, Jingjing Wang, John C Vederas, Yi Tang
PMID22406519
ABSTRACT
Iterative highly reducing polyketide synthases from filamentous fungi are the most complex and enigmatic type of polyketide synthase discovered to date. Here we uncover an unusual degree of programming by the hypothemycin highly reducing polyketide synthase, in which a single ketoreductase domain shows stereospecificity that is controlled by substrate length. Mapping of the structural domains responsible for this feature allowed for the biosynthesis of an unnatural diastereomer of the natural product dehydrozearalenol.