- Albendazole sulfoxide enantiomers: preparative chiral separation and absolute stereochemistry.
Albendazole sulfoxide enantiomers: preparative chiral separation and absolute stereochemistry.
Journal of chromatography. A (2012-02-22)
Tiago C Lourenço, João M Batista, Maysa Furlan, Yanan He, Laurence A Nafie, Cesar C Santana, Quezia B Cass
PMID22341660
ABSTRACT
The enantiomeric separation of albendazole sulfoxide was carried out by simulated moving bed chromatography with variable zones (VARICOL). An overall recovery of 97% was achieved and enantiomeric ratios of 99.5% for raffinate and 99.0% for extract were attained. A total of 880 mg of (+)-albendazol sulfoxide and 930 mg of its antipode were collected after 55 cycles or 11 h of process, resulting in a mass rate of 2 g/day. Furthermore the absolute configuration of the enantiopure compounds was determined for the first time by vibrational circular dichroism (VCD) with the aid of theoretical calculations as (-)-(S) and (+)-(R)-albendazole sulfoxide.