Merck
  • Cytokine production by infrapatellar fat pad can be stimulated by interleukin 1β and inhibited by peroxisome proliferator activated receptor α agonist.
Cytokine production by infrapatellar fat pad can be stimulated by interleukin 1β and inhibited by peroxisome proliferator activated receptor α agonist.
Annals of the rheumatic diseases (2012-02-07)
Stefan Clockaerts, Yvonne M Bastiaansen-Jenniskens, Carola Feijt, Luc De Clerck, J A N Verhaar, Anne-Marie Zuurmond, Vedrana Stojanovic-Susulic, Johan Somville, Margreet Kloppenburg, Gerjo J V M van Osch
ABSTRACT
Infrapatellar fat pad (IPFP) might be involved in osteoarthritis (OA) by production of cytokines. It was hypothesised that production of cytokines is sensitive to environmental conditions. To evaluate cytokine production by IPFP in response to interleukin (IL)1β and investigate the ability to modulate this response with an agonist for peroxisome proliferator activated receptor α (PPARα), which is also activated by lipid-lowering drugs such as fibrates. Cytokine secretion of IPFP was analysed in the medium of explant cultures of 29 osteoarthritic patients. IPFP (five donors) and synovium (six donors) were cultured with IL-1β and PPARα agonist Wy14643. Gene expression of IL-1β, monocyte chemoattractant protein (MCP1), (IL-6, tumour necrosis factor (TNF)α, leptin, vascular endothelial growth factor (VEGF), IL-10, prostaglandin-endoperoxide synthase (PTGS)2 and release of TNFα, MCP1 and prostaglandin E(2) were compared with unstimulated IPFP and synovium explants. IPFP released large amounts of inflammatory cytokines, adipokines and growth factors. IL-1β increased gene expression of PTGS2, TNFα, IL-1β, IL-6 and VEGF and increased TNFα release in IPFP. MCP1, leptin, IL-10 gene expression and MCP1, leptin and PGE(2) release did not increase significantly. Synovium responded to IL-1β similarly to IPFP, except for VEGF gene expression. Wy14643 decreased gene expression of PTGS2, IL-1β, TNFα, MCP1, VEGF and leptin in IPFP explants and IL-1β, TNFα, IL-6, IL-10 and VEGF in synovium that responded to IL-1β. IPFP is an active tissue within the joint. IPFP cytokine production is increased by IL-1β and decreased by a PPARα agonist. The effects were similar to effects seen in synovium. Fibrates may represent a potential disease-modifying drug for OA by modulating inflammatory properties of IPFP and synovium.