- Mukaiyama-Michael reactions with acrolein and methacrolein: a catalytic enantioselective synthesis of the C17-C28 fragment of pectenotoxins.
Mukaiyama-Michael reactions with acrolein and methacrolein: a catalytic enantioselective synthesis of the C17-C28 fragment of pectenotoxins.
Organic letters (2012-02-03)
Eeva K Kemppainen, Gokarneswar Sahoo, Arto Valkonen, Petri M Pihko
PMID22296172
ABSTRACT
Enantioselective iminium-catalyzed reactions with acrolein and methacrolein are rare. A catalytic enantioselective Mukaiyama-Michael reaction that readily accepts acrolein or methacrolein as substrates, affording the products in good yields and 91-97% ee, is presented. As an application of the methodology, an enantioselective route to the key C17-C28 segment of the pectenotoxin using the Mukaiyama-Michael reaction as the key step is described.