Merck
D-pinitol inhibits RANKL-induced osteoclastogenesis.
International immunopharmacology (2012-01-25)
Shan-Chi Liu, Show-Mei Chuang, Chih-Hsin Tang
ABSTRACT
Numerous studies have indicated that inflammatory cytokines play a major role in osteoclastogenesis, leading to the bone resorption that is frequently associated with osteoporosis. D-pinitol, a 3-methoxy analogue of D-chiroinositol, was identified as an active principle in soy foods and legumes. Here we found that D-pinitol markedly inhibited the receptor activator of nuclear factor kappa B ligand (RANKL)-induced osteoclastic differentiation from bone marrow stromal cells and RAW264.7 macrophage cells. In addition, D-pinitol also reduced RANKL-induced p38 and JNK phosphorylation. Furthermore, RANKL-mediated increase of IKK, IκBα, and p65 phosphorylation and NF-κB-luciferase activity was inhibited by D-pinitol. However, D-pinitol did not affect the proliferation and differentiation of osteoblasts. In addition, D-pinitol also prevented the bone loss induced by ovariectomy in vivo. Our data suggest that D-pinitol inhibits osteoclastogenesis from bone marrow stromal cells and macrophage cells via attenuated RANKL-induced p38, JNK, and NF-κB activation, which in turn protect bone loss from ovariectomy.