Merck
  • A coordination polymer nanobelt (CPNB)-based aptasensor for sulfadimethoxine.
A coordination polymer nanobelt (CPNB)-based aptasensor for sulfadimethoxine.
Biosensors & bioelectronics (2012-01-17)
Kyung-Mi Song, Euiyoung Jeong, Weejeong Jeon, Hunho Jo, Changill Ban
ABSTRACT
A polymer-based aptasensor, which consisted of fluorescein amidite (FAM)-modified aptamers and coordination polymer nanobelts (CPNBs), was developed utilizing the fluorescence quenching effect to detect sulfadimethoxine residue in food products. A single-stranded DNA (ssDNA) aptamer, which was a specific bio-probe for sulfadimethoxine (Su13; 5'-GAGGGCAACGAGTGTTTATAGA-3'), was discovered by a magnetic bead-based systematic evolution of ligands by exponential enrichment (SELEX) technique, and the fluorescent quenchers CPNBs were produced by mixing AgNO(3) and 4,4'-bipyridine. This aptasensor easily and sensitively detected sulfadimethoxine in solution with a limit of detection (LOD) of 10ng/mL. Furthermore, the antibiotic dissolved in milk was also effectively detected with the same LOD value. In addition, this aptamer probe offered high specificity for sulfadimethoxine compared to other antibiotics. These valuable results provide ample evidence that the CPNB-based aptasensor can be used to quantify sulfadimethoxine residue in food products.
Product Description
Supelco
Sulfadimethoxine, 98.0-102.0%