Merck
  • Photocurrent and electronic activities of oriented-His-tagged photosynthetic light-harvesting/reaction center core complexes assembled onto a gold electrode.
Photocurrent and electronic activities of oriented-His-tagged photosynthetic light-harvesting/reaction center core complexes assembled onto a gold electrode.
Biomacromolecules (2012-01-14)
Masaharu Kondo, Kouji Iida, Takehisa Dewa, Hirofumi Tanaka, Takuji Ogawa, Sakiko Nagashima, Kenji V P Nagashima, Keizo Shimada, Hideki Hashimoto, Alastair T Gardiner, Richard J Cogdell, Mamoru Nango
ABSTRACT
A polyhistidine (His) tag was fused to the C- or N-terminus of the light-harvesting (LH1)-α chain of the photosynthetic antenna core complex (LH1-RC) from Rhodobacter sphaeroides to allow immobilization of the complex on a solid substrate with defined orientation. His-tagged LH1-RCs were adsorbed onto a gold electrode modified with Ni-NTA. The LH1-RC with the C-terminal His-tag (C-His LH1-RC) on the modified electrode produced a photovoltaic response upon illumination. Electron transfer is unidirectional within the RC and starts when the bacteriochlorophyll a dimer in the RC is activated by light absorbed by LH1. The LH1-RC with the N-terminal His-tag (N-His LH1-RC) produced very little or no photocurrent upon illumination at any wavelength. The conductivity of the His-tagged LH1-RC was measured with point-contact current imaging atomic force microscopy, indicating that 60% of the C-His LH1-RC are correctly oriented (N-His 63%). The oriented C-His LH1-RC or N-His LH1-RC showed semiconductive behavior, that is, had the opposite orientation. These results indicate that the His-tag successfully controlled the orientation of the RC on the solid substrate, and that the RC produced photocurrent depending upon the orientation on the electrode.