- Synthesis of novel D-ring fused 7'-aryl-androstano[17,16-d][1,2,4] triazolo[1,5-a]pyrimidines.
Synthesis of novel D-ring fused 7'-aryl-androstano[17,16-d][1,2,4] triazolo[1,5-a]pyrimidines.
Steroids (2011-12-21)
Li-Hua Huang, Yong-Fei Zheng, Chuan-Jun Song, Yan-Guang Wang, Zhi-Yu Xie, Yao-Wen Lai, Yong-Zheng Lu, Hong-Min Liu
PMID22182831
ABSTRACT
The preparation of novel steroidal heterocycles containing the 7-aryl-substituted 1,2,4-triazolo[1,5-a]pyrimidine moiety fused to the 16,17-positions of the steroid nucleus is described. The Aldol reaction of 4-aza-androst-3,17-dione (1a) and dehydroepiandrosterone (DHEA, 1b) with aromatic aldehydes was catalyzed by KF/Al(2)O(3) to give the corresponding 3-oxo-4-aza-5α- and 3β-hydroxy-5-en-16-arylidene-17-ketosteroids (2a-r). Subsequently, the intermediates 2a-r reacted with dinucleophilic 3-amino-1,2,4-triazole in presence of t-BuOK to afford the title compounds (3a-r). All the synthesized heterosteroids are new and are currently being evaluated for their biological activities.