Merck
  • DDTs in rice frogs (Rana limnocharis) from an agricultural site, South China: tissue distribution, biomagnification, and potential toxic effects assessment.
DDTs in rice frogs (Rana limnocharis) from an agricultural site, South China: tissue distribution, biomagnification, and potential toxic effects assessment.
Environmental toxicology and chemistry (2011-12-16)
Jiang-Ping Wu, Ying Zhang, Xiao-Jun Luo, She-Jun Chen, Bi-Xian Mai
ABSTRACT
Contamination with agricultural pesticides such as dichlorodiphenyltrichloroethane (DDT) and its metabolites, dichlorodiphenyldichloroethylene (DDE) and dichlorodiphenyldichloroethane (DDD), is among several proposed stressors contributing to the global declines in amphibian populations and species biodiversity. These chemicals were examined in insects and in the muscle, liver, and eggs of rice frogs (Rana limnocharis) from the paddy fields of an agricultural site in South China. The ΣDDT (sum of DDT, DDE, and DDD) concentrations ranged from 154 to 915, 195 to 1,400, and 165 to 1,930 ng/g lipid weight in the muscle, liver, and eggs, respectively. All the DDTs (DDT, DDE, and DDD) showed higher affinity for the liver relative to muscle tissue and can be maternally transferred to eggs in female frogs. The average biomagnification factors for DDTs ranged from 1.6 to 1.9 and 1.5 to 2.9 in female and male frogs, respectively, providing clear evidence of their biomagnification from insects to frogs. Compared with the reported DDT levels demonstrated to have toxic effects on frogs, DDTs in the present frogs are unlikely to constitute an immediate health risk. However, the adverse impacts of high DDT residues in eggs on the hatching success and their potential toxicity to the newly metamorphosed larval frogs should be assessed further.