Merck
  • Synergistic antibacterial and antibiotic effects of bisbenzylisoquinoline alkaloids on clinical isolates of methicillin-resistant Staphylococcus aureus (MRSA).
Synergistic antibacterial and antibiotic effects of bisbenzylisoquinoline alkaloids on clinical isolates of methicillin-resistant Staphylococcus aureus (MRSA).
Molecules (Basel, Switzerland) (2011-11-26)
Guo-Ying Zuo, Yang Li, Tao Wang, Jun Han, Gen-Chun Wang, Yun-Ling Zhang, Wei-Dong Pan
ABSTRACT
The antibacterial activity of two bisbenzylisoquinoline alkaloids, tetrandrine (Tet) and demethyltetrandrine (d-Tet), alone and in combination with the antibiotics ampicillin (AMP), azithromycin (AZM), cefazolin (CFZ) and levofloxacin (LEV) against 10 clinical isolates of staphylococcal chromosomal cassette mec (SCCmec) III type methicillin-resistant Staphylococcus aureus (MRSA) was studied. Susceptibility to each agent alone was tested using a broth microdilution method. The chequerboard and time-kill tests were used for the combined evaluations. The minimal inhibitory concentrations/minimal bactericidal concentrations (MICs/MBCs, μg/mL) ranges alone were 64-128/256-1,024 for both Tet and d-Tet. Significant synergies against 90% of the isolates were observed for the Tet/CFZ combination, with their MICs being reduced by 75-94% [fractional inhibitory concentration indices (FICIs) ranged from 0.188 to 0.625], respectively. An additive bactericidal result was also observed for the Tet (d-Tet)/CFZ combination in the time-kill experiments. These results demonstrated that Tet and d-Tet enhanced the in vitro inhibitory efficacy of CFZ. Their potential for combinatory therapy of patients infected with MRSA warrants further pharmacological investigation.