Merck
  • Capreomycin--a polypeptide antitubercular antibiotic with unusual binding properties toward copper(II).
Capreomycin--a polypeptide antitubercular antibiotic with unusual binding properties toward copper(II).
Journal of inorganic biochemistry (2011-11-25)
Kamila Stokowa, Wojciech Szczepanik, Nicola Gaggelli, Elena Gaggelli, Gianni Valensin, Małgorzata Jeżowska-Bojczuk
ABSTRACT
Capreomycin is an important therapeutic agent having intriguing and diverse molecular features. Its polypeptidic structure rich in nitrogen donors makes the drug a promising chelating agent for a number of transition metal ions, especially for copper(II). The results of the model investigational studies suggest that capreomycin anchors Cu(2+) ion with an amino function of the α,β-diaminopropionic acid residue at pH around 5. At physiological pH copper(II) ion is coordinated by two deprotonated amide nitrogen atoms of the α,β-diaminopropionic acid, the serine residue as well as the amino function deriving from the β-lysine. Above that pH value we observe a rearrangement within the coordination sphere leading to movement of Cu(2+) to the center of the peptide ring with concurrent coordination of four nitrogen donors. Spin-lattice relaxation enhancements and potentiometric measurements clearly indicate that deprotonated amide nitrogen atom from the β-ureidodehydroalanine moiety is the fourth donor atom.