- Identification of a new 80 k isoform of phosphatidylinositol 4-phosphate 5-kinase from bovine brain.
Identification of a new 80 k isoform of phosphatidylinositol 4-phosphate 5-kinase from bovine brain.
FEBS letters (1993-07-05)
M Schmidt, M Varsányi, R Thieleczek, L M Heilmeyer
PMID8391476
ABSTRACT
Phosphatidylinositol 4-phosphate 5-kinase is associated with bovine brain microsomes to an extent of approximately 65% of the total cellular enzyme activity. This membrane-associated kinase activity can be solubilized with Triton X-114. Following polyacrylamide gel electrophoresis in the presence of SDS the enzyme can be renaturated from gel slices in the presence of desoxycholate and Triton X-100. Catalytic activity appears at an apparent molecular weight of 80 k. Analysis of the reaction product formed by the 80 k protein reveals the existence of a 5-phosphotransferase, identifying the 80 k polypeptide as a new phosphatidylinositol 4-phosphate 5-kinase isoform.