Merck
  • Asiaticoside: attenuation of neurotoxicity induced by MPTP in a rat model of Parkinsonism via maintaining redox balance and up-regulating the ratio of Bcl-2/Bax.
Asiaticoside: attenuation of neurotoxicity induced by MPTP in a rat model of Parkinsonism via maintaining redox balance and up-regulating the ratio of Bcl-2/Bax.
Pharmacology, biochemistry, and behavior (2011-10-18)
Chang-Liang Xu, Qi-Zhi Wang, Ling-Mei Sun, Xiu-Min Li, Ji-Min Deng, Lu-Fan Li, Jin Zhang, Rong Xu, Shi-Ping Ma
ABSTRACT
In this study, we investigated the neuroprotective effects of asiaticoside, a triterpenoid saponin isolated from the Chinese medicinal herb Centella asiatica, in the rats model of Parkinsonism induced by 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP). Rats were first injected with MPTP. One day after surgery, asiaticoside was administered and the behavioral tests were assessed. On 14th day, the rats were sacrificed, substantia nigra (SN) and striatum were dissected, and then dopamine (DA) and its metabolites in striatum and malonyldialdehyde (MDA) contents, reduced glutathione (GSH) level and gene expression level in SN were estimated. Treatment with asiaticoside was found to protect dopaminergic neuron by antagonizing MPTP induced neurotoxicity and to improve locomotor dysfunction. Asiaticoside significantly attenuated the MPTP-induced reduction of dopamine in the striatum. The content of MDA was significantly decreased while the GSH level was significantly increased in asiaticoside-treated groups. In addition, asiaticoside increased the Bcl-2/Bax ratio. These results indicated that asiaticoside was effective in reversing MPTP induced Parkinsonism via its neuroprotective effects including antioxidant activity, maintaining the metabolic balance of DA, and increasing ratio of Bcl-2/Bax.