Merck
Three dye energy transfer cascade within DNA thin films.
Chemical communications (Cambridge, England) (2011-10-15)
Daminda Navarathne, Yogesh Ner, James G Grote, Gregory A Sotzing
ABSTRACT
An efficient cascade FRET was realized in solid state DNA-CTMA thin films using a three chromophore system without any covalent attachments. The extent of energy transfer from Cm102 to SRh was studied and found to improve eight-fold using the bridging dye Pm567.