- Synthesis and anti-nociceptive and anti-inflammatory effects of gaultherin and its analogs.
Synthesis and anti-nociceptive and anti-inflammatory effects of gaultherin and its analogs.
Journal of Asian natural products research (2011-08-13)
Chao Wang, Tian-Tai Zhang, Guan-Hua Du, Dong-Ming Zhang
PMID21830886
ABSTRACT
The synthesis of gaultherin (1) and its analogs was carried out to provide 11 glycosides under phase-transfer catalytic conditions. The activities of all synthesized compounds were evaluated by nitric oxide production inhibitory assay in vitro. Methyl 2-O-(4-O-β-d-galactopyranosyl)-β-d-glucopyranosylbenzoate (5f) showed significantly anti-nociceptive and anti-inflammatory effects by the evaluation in vivo. Structure-activity relationships within these compounds were discussed.