- [Study on the chemical constituents from Melicope ptelefolia].
[Study on the chemical constituents from Melicope ptelefolia].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2011-08-10)
Yu-Feng Xie, Yue Liang, Qing-Tao Du, Li-Bing Guo
PMID21823455
ABSTRACT
To study the chemical constituents from Melicope ptelefolia. Several chromatographic methods were applied to isolate and purify compounds. Their structures were identified on the basis of physicochemical properties and spectroscopic data. Seven compounds were isolated and elucidated as n-octadecanyl palmitate (I), beta-sitosterol (II), palmitic acid (III), 3, 5,3'-trihydroxy-8,4'-dimethoxy-7-(3-methylbut-2-enyloxy) flavone (IV), daucosterol (V), salylic acid (VI), kaempferol-3-O-alpha-D-arabinpyranoside (VII). Compound VII is isolated from the genus for the first time, Compounds V and VI are isolated from Melicope ptelefolia for the first time.