Merck
  • Synthesis, structures, cellular uptake and apoptosis-inducing properties of highly cytotoxic ruthenium-Norharman complexes.
Synthesis, structures, cellular uptake and apoptosis-inducing properties of highly cytotoxic ruthenium-Norharman complexes.
Dalton transactions (Cambridge, England : 2003) (2011-08-02)
Caiping Tan, Shouhai Wu, Sensen Lai, Minxu Wang, Yu Chen, Lingjun Zhou, Yiping Zhu, Wu Lian, Wenlie Peng, Liangnian Ji, Anlong Xu
ABSTRACT
Three novel Ru(II) complexes of the general formula [Ru(N-N)(2)(Norharman)(2)](SO(3)CF(3))(2), where N-N = 2,2'-bipyridine (bpy, 1), 1,10-phenanthroline (phen, 2), 4,7-diphenyl-1,10-phenanthroline (DIP, 3) and Norharman (9H-pyrido[3,4-b]indole) is a naturally occurring β-carboline alkaloid, have been synthesized and characterized. The molecular structures of 1 and 2 have been determined by X-ray diffraction analysis. The cellular uptake efficiencies, in vitro cytotoxicities and apoptosis-inducing properties of these complexes have been extensively explored. Notably, 1-3 exhibit potent antiproliferative activities against a panel of human cancer cell lines with IC(50) values lower than those of cisplatin. Further studies show that 1-3 can cause cell cycle arrest in the G0/G1 phase and induce apoptosis through mitochondrial dysfunction and reactive oxygen species (ROS) generation. In vitro DNA binding studies have also been conducted to provide information about the possible mechanism of action.