Merck
β-Olefination of 2-alkynoates leading to trisubstituted 1,3-dienes.
Organic letters (2011-06-09)
Mathias J Jacobsen, Erik Daa Funder, Jacob R Cramer, Kurt V Gothelf
ABSTRACT
A phosphine-mediated olefination of 2-alkynoates with aldehydes forming 1,3-dienes with high E-selectivity and up to 88% yield is described. Reaction conditions are optimized and reactions are demonstrated for various aryl, alkyl, and alkenyl aldehydes and for ethyl 2-alkynoates with different substituents in the δ-position. Proof of concept is shown for the generation of a β,γ-unsaturated lactone by intramolecular olefination, and furthermore the use of the generated 1,3-dienes in the Diels-Alder reaction has been demonstrated.