Merck
  • Concentrations, distribution, and bioaccumulation of synthetic musks in the Haihe River of China.
Concentrations, distribution, and bioaccumulation of synthetic musks in the Haihe River of China.
Chemosphere (2011-06-01)
Zhengjun Hu, Yali Shi, Yaqi Cai
ABSTRACT
Seven typical synthetic musks (SMs) in the samples from the surface water, sediment and fish of the Haihe River were measured. The SM concentrations in the sediment and surface water of the Haihe River were significantly lower than those in the Dagu Drainage River and Chentaizi Drainage River (p<0.05). Along the flow direction, the SM concentrations in surface water and sediment tended to increase from the upstream to the downstream of Dagu Drainage River. The Bioaccumulation factors (BAFs) of galaxolide (HHCB) and tonalide (AHTN) were calculated at high levels in the muscles of crucian carp, common carp, and silver carp. Most of the biota-sediment accumulation factors (BSAFs) for HHCB and AHTN were higher than 1.7, suggesting magnification possibly exist in the musk bioaccumulations of the three fishes in the Haihe River. No significant differences in HHCB/AHTN ratios were observed among the water, fish, and sediment samples (p>0.05). However, the HHCB/AHTN values in the Haihe River were much lower than those in the Dagu Drainage River and Chentaizi Drainage River (p<0.05). Compared with several typical persistent organic pollutants (POPs), the musk concentrations were higher or comparable in the Haihe River.
Sigma-Aldrich
6-Acetyl-1,1,2,4,4,7-hexamethyltetralin, ≥98%