Merck
  • Total synthesis of optically active lycopladine A by utilizing diastereoselective protection of carbonyl group in a 1,3-cyclohexanedione derivative.
Total synthesis of optically active lycopladine A by utilizing diastereoselective protection of carbonyl group in a 1,3-cyclohexanedione derivative.
The Journal of organic chemistry (2011-05-06)
Kou Hiroya, Yoshihiro Suwa, Yusuke Ichihashi, Kiyofumi Inamoto, Takayuki Doi
ABSTRACT
We successfully synthesized two enantiomers of bicyclic enones, (7R,7aR)- and (7S,7aS)-9, from the hemiacetal 2a, which we first synthesized from the symmetrical diketone 1a via diastereoselective carbon-oxygen bond formation between one of the carbonyl groups and the chiral alcohol on the C2 side chain in a 2,2-disubstituted 1,3-cycloalkanedione derivative. We also report the total synthesis of natural (+)-lycopladine A [(+)-6] from (7R,7aR)-9 and the formal synthesis of unnatural (-)-lycopladine A [(-)-6] from (7S,7aS)-9.