Merck
  • Cellular and molecular actions displayed by estrone on vascular endothelium.
Cellular and molecular actions displayed by estrone on vascular endothelium.
Molecular and cellular endocrinology (2011-05-04)
M Belén Rauschemberger, Marisa J Sandoval, Virginia L Massheimer
ABSTRACT
In this work we provide evidence that estrone "per se" modulates cellular endothelial growth and survival, events that play key roles in the development of vascular disease. Moreover, under oxidative stress conditions the hormone prevented apoptosis triggered by hydrogen peroxide. Although estrone did not affect E-selectin and VCAM-1 mRNAs synthesis, the hormone prevented the expression of these adhesion molecules induced by the proinflammatory agent LPS. The steroid partially attenuated leukocyte adhesion not only under basal conditions but also in the presence of LPS. Using ICI182780 compound as estrogen receptor antagonist, and PD98059 as MAPK inhibitor we obtained evidence that the mitogenic action of estrone involved the participation of ER and MAPK transduction pathway activation. The presence of estradiol impaired the effect of estrone on cell proliferation and vasoactive production. These results suggest that estrone exhibits a remarkable biological action on endothelial cells, modulating vasoactive production, proliferation, apoptosis, and cell adhesion events.