Merck
  • Determination of betaines in vegetable oils by capillary electrophoresis tandem mass spectrometry--application to the detection of olive oil adulteration with seed oils.
Determination of betaines in vegetable oils by capillary electrophoresis tandem mass spectrometry--application to the detection of olive oil adulteration with seed oils.
Electrophoresis (2011-04-27)
Laura Sánchez-Hernández, María Castro-Puyana, María Luisa Marina, Antonio L Crego
ABSTRACT
A CE-tandem mass spectrometry (MS²) methodology enabling the simultaneous determination of betaines (glycine betaine, trigonelline, proline betaine and total content of carnitines) in vegetable oils was developed. Betaines were derivatized with butanol previous to their baseline separation in 10 min using a 0.1 M formic acid buffer at pH 2.0. Ion trap conditions were optimized in order to maximize the selectivity and sensitivity. Analytical characteristics of the proposed method were established by evaluating its selectivity, linearity, precision (RSDs ranged from 4.8 to 10.7% for corrected peak areas) and accuracy by means of recovery studies (from 80 to 99%) and LODs and LOQs at 0.1 ppb level. The method was applied for the determination of the selected betaines in seed oils and extra virgin olive oils. MS² experiments provided the fingerprint fragmentation for the betaines identified in vegetable oils. In extra virgin olive oils, carnitines were not detected, making it possible to propose them as a feasible novel marker for the detection of adulterations of olive oils. Application of the developed method for the analysis of different mixtures of extra virgin olive oil with seed oil (between 2 and 10%) enabled the detection and quantitation of the total content of carnitines. The results obtained show the high potential of the developed method for the authentication and quality control of olive oils.