- Efficacy of rufinamide in drug-resistant epilepsy: a meta-analysis.
Efficacy of rufinamide in drug-resistant epilepsy: a meta-analysis.
Pediatric neurology (2011-04-13)
Alberto Verrotti, Giulia Loiacono, Enzo Ballone, Peter A Mattei, Francesco Chiarelli, Paolo Curatolo
PMID21481742
ABSTRACT
Rufinamide is a new orally active antiepileptic drug that has been found to be effective in the treatment of partial seizures and drop attacks associated with Lennox-Gastaut syndrome. We performed a quantitative analysis of the efficacy of this new antiepileptic drug from all double-blind, add-on, randomized, placebo-controlled clinical trials published to date. Data from 918 patients were studied. The number of patients per study varied from 25 to 262. Rufinamide was efficacious in doses up to 45 mg/kg daily when provided as adjunctive therapy in patients with Lennox-Gastaut syndrome and other drug-resistant epilepsies. Further studies are needed to confirm and expand these findings.