- Akt/mammalian target of rapamycin signaling pathway regulates neurite outgrowth in cerebellar granule neurons stimulated by methylcobalamin.
Akt/mammalian target of rapamycin signaling pathway regulates neurite outgrowth in cerebellar granule neurons stimulated by methylcobalamin.
Neuroscience letters (2011-04-05)
Kiyoshi Okada, Hiroyuki Tanaka, Ko Temporin, Michio Okamoto, Yusuke Kuroda, Hisao Moritomo, Tsuyoshi Murase, Hideki Yoshikawa
PMID21458538
ABSTRACT
Methylcobalamin (MeCbl), a vitamin B12 analog, promotes neurite outgrowth by activating Akt in neurons. However, Akt is involved in many cellular functions, and the downstream signal of Akt that promotes neurite outgrowth in neurons in the presence of MeCbl remains obscure. Mammalian target of rapamycin (mTOR) is a serine/threonine protein kinase that regulates multiple cellular functions including neurite outgrowth. mTOR is regarded as important for the regeneration of injured nerves. In this study, we examined the relationship between MeCbl and mTOR activity and found that MeCbl increases mTOR activity via the activation of Akt and promotes neurite outgrowth in cerebellar granule neurons via the activation of mTOR.