Merck
Direct conversion of benzylic and allylic alcohols to phosphonates.
The Journal of organic chemistry (2011-03-17)
Rocky J Barney, Rebekah M Richardson, David F Wiemer
ABSTRACT
Benzyl phosphonate esters often serve as reagents in Horner-Wadsworth-Emmons reactions. In most cases, they can be prepared from benzylic alcohols via formation of the corresponding halide followed by an Arbuzov reaction. To identify a more direct synthesis of phosphonate esters, we have developed a one-flask procedure for conversion of benzylic and allylic alcohols to the corresponding phosphonates through treatment with triethyl phosphite and ZnI(2).