Merck
  • Influences of treatment of early pregnant mares with the progestin altrenogest on embryonic development and gene expression in the endometrium and conceptus.
Influences of treatment of early pregnant mares with the progestin altrenogest on embryonic development and gene expression in the endometrium and conceptus.
Theriogenology (2011-03-15)
C Willmann, S Budik, I Walter, C Aurich
ABSTRACT
A positive influence of altrenogest treatment on a retarded development of the conceptus around the beginning of placentation in mares older than 8 years could be recently demonstrated. In the present study, effects of altrenogest treatment in early-pregnant mares on conceptus development and expression of endometrial and embryonic genes were investigated. Genes were chosen according to a possible involvement in embryo-maternal interaction and embryonic development in the equine species. Mares were treated with altrenogest (0.044 mg/kg bodyweight) or sunflower oil (placebo) from day 5 to 11 after ovulation. Embryos (altrenogest n = 13, placebo n = 12) and biopsies were collected on day 11. Pregnancy rate and embryonic size were not influenced by treatment (embryonic diameter: altrenogest 7.0 ± 2.5, placebo 6.5 ± 1.7 mm, n.s.). The percentage of luminal epithelial cells, superficial glandular epithelial cells and interstitial cells with nuclei staining positively for the progesterone receptor was significantly lower (P < 0.05) in samples collected from altrenogest-treated than from placebo-treated mares (e.g., luminal epithelium: altrenogest 1.9 ± 1.7%, placebo 23.0 ± 10.5%, P < 0.05). Staining for COX2 (cyclooxygenase-2) was not affected by treatment. In the endometrium a slight but significant increase in the number of PMN (polymorph nuclear neutrophils) was seen in response to treatment (altrenogest 0.8 ± 0.5 PMN/field, placebo 0.3 ± 0.3 PMN/field; P < 0.05). No differences in the relative gene expression of COX2, the receptors for progesterone, estrogens and growth hormone as well as for IGF (insulin-like growth factor) 1 and 2 were detected. The relative gene expression of aquaporin 3 in relation to β-actin differed significantly (P < 0.05) between embryos from altrenogest (3.2 ± 0.8) and placebo-treated mares (1.3 ± 0.2), but no other genes were affected. The study demonstrates down-regulation of progesterone receptors in the endometrium of early pregnant mares by treatment with the progestin altrenogest. This increased expression of aquaporin 3 in the conceptus was not related to changes in embryonic size or development.