Merck
  • Mechanisms of drug sensitization to TRA-8, an agonistic death receptor 5 antibody, involve modulation of the intrinsic apoptotic pathway in human breast cancer cells.
Mechanisms of drug sensitization to TRA-8, an agonistic death receptor 5 antibody, involve modulation of the intrinsic apoptotic pathway in human breast cancer cells.
Molecular cancer research : MCR (2011-03-02)
Hope M Amm, Tong Zhou, Adam D Steg, Huichien Kuo, Yufeng Li, Donald J Buchsbaum
ABSTRACT
TRA-8, a monoclonal antibody to death receptor 5 induces apoptosis in various cancer cells; however, the degree of sensitivity varies from highly sensitive to resistant. We have previously shown that resistance to TRA-8 can be reversed by using chemotherapeutic agents, but the mechanism underlying this sensitization was not fully understood. Here, we examined the combination of TRA-8 with doxorubicin or bortezomib in breast cancer cells. In TRA-8-resistant BT-474 and T47D cells, both chemotherapy agents synergistically sensitized cells to TRA-8 cytotoxicity with enhanced activation of apoptosis shown by cleavage of caspases and PARP, reduced Bid, increased proapoptotic Bcl-2 proteins, and increased mitochondrial membrane depolarization. Doxorubicin or bortezomib combined with TRA-8 also reduced Bcl-XL and X-linked inhibitors of apoptosis (XIAP) in treated cells. Furthermore, targeting these proteins with pharmacologic modulators, AT-101, BH3I-2' and AT-406, produced sensitization to TRA-8. TRA-8 combined with AT-101 or BH3I-2', inhibitors of antiapoptotic Bcl-2 proteins, produced synergistic cytotoxicity against ZR-75-1, BT-474, and T47D cells. The IAP-targeting compound, AT-406, was synergistic with TRA-8 in BT-474 cells, and to a lesser extent T47D cells. Activation of the intrinsic apoptotic pathway was a common mechanism associated with sensitization of TRA-8-resistant breast cancer cell lines. Collectively, these studies show that the Bcl-2 and IAP families of proteins are involved in TRA-8 and chemotherapy resistance via their modulation of the intrinsic apoptotic pathway. Targeting these proteins with novel agents sensitized TRA-8-resistant breast cancer cells, suggesting this approach may represent a potent therapeutic strategy in the treatment of breast cancer.