Merck
  • Investigating wound healing, tyrosinase inhibitory and antioxidant activities of the ethanol extracts of Salvia cryptantha and Salvia cyanescens using in vivo and in vitro experimental models.
Investigating wound healing, tyrosinase inhibitory and antioxidant activities of the ethanol extracts of Salvia cryptantha and Salvia cyanescens using in vivo and in vitro experimental models.
Journal of ethnopharmacology (2011-03-01)
Ipek Süntar, Esra Küpeli Akkol, Fatma Sezer Senol, Hikmet Keles, Ilkay Erdogan Orhan
ABSTRACT
Salvia L. species are widely used against wounds and skin infections in Turkish folk medicine. The aim of the present study is to evaluate wound healing activity of the ethanol (EtOH) extracts of Salvia cryptantha and Salvia cyanescens. For the assessment of wound healing activity linear incision and circular excision wound models were employed on rats and mice. The wound healing effect was comparatively evaluated with the standard skin ointment Madecassol(®). Inhibition of tyrosinase, a key enzyme in skin aging, was achieved using ELISA microplate reader. Antioxidant activity was evaluated by 2,2-diphenyl-1-picrylhydrazyl (DPPH) and superoxide radical scavenger effect, ferrous ion-chelating ability, and ferric-reducing antioxidant power (FRAP) tests. The EtOH extract of Salvia cryptantha treated groups of animals showed 56.5% contraction, whereas the reference drug Madecassol(®) showed 100% contraction. On the other hand, the same extract on linear incision wound model demonstrated a significant increase (33.2%) in wound tensile strength as compared to other groups. The results of histopathological examination maintained the upshot of linear incision and circular excision wound models as well. These findings specify that Salvia cryptantha for wound healing activity can be appealed further phytochemical estimation for spotting its active components.