Merck
  • [Effect of m-nisoldipine on the Ca2+/CaM/CaN signal pathway in 5-HT-induced proliferation of rat PASMCs].
[Effect of m-nisoldipine on the Ca2+/CaM/CaN signal pathway in 5-HT-induced proliferation of rat PASMCs].
Yao xue xue bao = Acta pharmaceutica Sinica (2011-03-01)
Xue-Yan Chen, Huan-Long Liu, Zhen-Hua Pan, Qing-Feng Miao, Yong-Jian Zhang
ABSTRACT
This study is to explore the activation of the Ca2+/CaM/CaN signal pathway in 5-HT-induced proliferation of rat pulmonary artery smooth muscle cells (PASMCs) and the inhibitory effect of m-nisoldipine (m-Nis) on this pathway. PASMCs were cultured with the explant technique. The proliferation of PASMCs was evaluated by MTT assay. Confocal microscopy was used to measure the change of [Ca2+]i. The mRNA expression of CaM and CaN was evaluated by RT-PCR and the activity of CaN was measured according to the instruction of kits. The results of MTT assay suggested that 5-HT (1 micromol x L(-1)) significantly induced the proliferation of rat PASMCs (P < 0.01), which was inhibited obviously by m-Nis (P < 0.05 or P < 0.01). Similarly, m-Nis inhibited 5-HT-induced elevation of [Ca2+]i (P < 0.01). The mRNA expression of CaM, CaN and the activation of CaN were also inhibited by m-Nis at different degrees (P < 0.05 or P < 0.01). Thus, the results of this study suggested that Ca2+/CaM/CaN signal pathway played an important role in 5-HT-induced proliferation of rat PASMCs, the inhibition of m-Nis on proliferation of rat PASMCs may be related to the blockage of Ca2+/CaM/CaN signal pathway by inhibiting the elevation of [Ca2+]i.