- A new pathway for poly(3-hydroxybutyrate) production in Escherichia coli and Corynebacterium glutamicum by functional expression of a new acetoacetyl-coenzyme A synthase.
A new pathway for poly(3-hydroxybutyrate) production in Escherichia coli and Corynebacterium glutamicum by functional expression of a new acetoacetyl-coenzyme A synthase.
Bioscience, biotechnology, and biochemistry (2011-02-11)
Ken'ichiro Matsumoto, Miwa Yamada, Chean Ring Leong, Sung-Jin Jo, Tomohisa Kuzuyama, Seiichi Taguchi
PMID21307588
ABSTRACT
A biosynthetic pathway for poly(3-hydroxybutyrate) [P(3HB)] was developed in Escherichia coli and Corynebacterium glutamicum by an acetoacetyl-coenzyme A (CoA) synthase (AACS) recently isolated from terpenoid-producing Streptomyces sp. strain CL190. Expression of AACS led to significant productions of P(3HB) in E. coli (10.5 wt %) and C. glutamicum (19.7 wt %).